Merck
  • Imbalanced unfolded protein response signaling contributes to 1-deoxysphingolipid retinal toxicity.
Imbalanced unfolded protein response signaling contributes to 1-deoxysphingolipid retinal toxicity.
Nature communications (2023-07-12)
Jessica D Rosarda, Sarah Giles, Sarah Harkins-Perry, Elizabeth A Mills, Martin Friedlander, R Luke Wiseman, Kevin T Eade
ABSTRACT
The accumulation of atypical, cytotoxic 1-deoxysphingolipids (1-dSLs) has been linked to retinal diseases such as diabetic retinopathy and Macular Telangiectasia Type 2. However, the molecular mechanisms by which 1-dSLs induce toxicity in retinal cells remain poorly understood. Here, we integrate bulk and single-nucleus RNA-sequencing to define biological pathways that modulate 1-dSL toxicity in human retinal organoids. Our results demonstrate that 1-dSLs differentially activate signaling arms of the unfolded protein response (UPR) in photoreceptor cells and Müller glia. Using a combination of pharmacologic activators and inhibitors, we show that sustained PERK signaling through the integrated stress response (ISR) and deficiencies in signaling through the protective ATF6 arm of the UPR are implicated in 1-dSL-induced photoreceptor toxicity. Further, we demonstrate that pharmacologic activation of ATF6 mitigates 1-dSL toxicity without impacting PERK/ISR signaling. Collectively, our results identify new opportunities to intervene in 1-dSL linked diseases through targeting different arms of the UPR.
Sigma-Aldrich
ISRIB, ≥98% (HPLC)