Skip to Content
Merck
All Photos(1)

Key Documents

407953

Sigma-Aldrich

Ionomycin, Calcium Salt, Streptomyces conglobatus in Solution

Useful in cell activation experiments when calcium dose-response data are not required. The solid form of this compound is also available.

Synonym(s):

SQ23377 calcium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

dry ice

storage temp.

−70°C

General description

Useful in cell activation experiments when calcium dose-response data are not required. The solid form of this compound (Cat. No. 407952) is also available.

Biochem/physiol Actions

Cell permeable: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (2 mg/268 µL) solution of Ionomycin, Calcium Salt, Streptomyces conglobatus (Cat. No. 407952) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C.

Other Notes

Dupuy d′Angeac, A., et al. 1993. J. Cell Physiol.154, 44.
Lee, H.C. 1993. J. Biol. Chem.268, 293.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meirong Li et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 129, 110425-110425 (2020-06-23)
Acute leukemia is a common hematological malignancy. Despite recent promising progress, the prognosis of acute leukemia patients remains to be improved. New therapies are therefore still needed. Salmonella typhimurium has been shown to be highly effective as an anti-tumor agent
Yuetong Wang et al.
Cell reports, 36(6), 109516-109516 (2021-08-12)
Although tumor-infiltrating lymphocytes (TILs) maintain their ability to proliferate, persist, and eradicate tumors, they are frequently dysfunctional in situ. By performing both whole-genome CRISPR and metabolic inhibitor screens, we identify that nicotinamide phosphoribosyltransferase (NAMPT) is required for T cell activation. NAMPT
Hongying Fu et al.
Nature communications, 11(1), 1050-1050 (2020-02-28)
Organisms respond to tissue damage through the upregulation of protective responses which restore tissue structure and metabolic function. Mitochondria are key sources of intracellular oxidative metabolic signals that maintain cellular homeostasis. Here we report that tissue and cellular wounding triggers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service