Skip to Content
Merck
All Photos(1)

Key Documents

407952

Sigma-Aldrich

Ionomycin, Calcium Salt, Streptomyces conglobatus

Ionomycin, Calcium Salt, Streptomyces conglobatus, CAS 56092-82-1, is useful in cell activation experiments when calcium dose-response data are not required.

Synonym(s):

Ionomycin, Calcium Salt, Streptomyces conglobatus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H70O9 · Ca
CAS Number:
Molecular Weight:
747.07
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C41H71O9.Ca/c1-25(21-29(5)34(43)24-35(44)30(6)22-27(3)20-26(2)14-15-38(46)47)12-11-13-28(4)39(48)31(7)36(45)23-33-16-18-41(10,49-33)37-17-19-40(9,50-37)32(8)42;/h11,13,24-33,36-37,39,42,45,48H,12,14-23H2,1-10H3,(H,46,47);/q-1;+2/p-1/b13-11+;/t25-,26-,27+,28-,29-,30+,31+,32-,33+,36+,37-,39-,40+,41+;/m1./s1

InChI key

RULWQOBWZOQCOY-SQNLDGQHSA-M

General description

Highly specific for divalent cations (Ca2+ > Mg2+ >> Sr2+ = Ba2+) and more effective than antibiotic A23187 (Cat. No. 100105) as a mobile ion carrier for Ca2+. Complexes with Ca2+ between pH 7 and 9.5, with intense UV absorption. In contrast to A23187, it is not fluorescent. Useful for studies of Ca2+ transport across biological membranes. Induces apoptotic neuronal degeneration in embryonic cortical neurons. Also induces cell cycle arrest in the G1 interval of mature Burkitt lymphoma cell lines. Often used in cell activation experiments when calcium dose-response data is not required.
Useful in cell activation experiments when calcium dose-response data are not required. Also available as a 10 mM solution in DMSO (Cat. No. 407953).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Useful in cell activation experiments
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Other Notes

Dupuy d′Angeac, A., et al. 1993. J. Cell Physiol.154, 44.
Lee, H.C. 1993. J. Biol. Chem.268, 293.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana P Saraiva et al.
Frontiers in oncology, 10, 1543-1543 (2020-09-26)
3D cell culture including different cell types, such as immune cells, is a representative platform that mimics the tumor microenvironment. Here we disclose an easy-to-handle 3D co-culture protocol using a scaffold-free technique with the breast cancer cell line MDA-MB-231 and
Jinfeng Cai et al.
eLife, 10 (2021-04-10)
The major barrier to curing HIV-1 infection is a small pool of latently infected cells that harbor replication-competent viruses, which are widely considered the origin of viral rebound when antiretroviral therapy (ART) is interrupted. The difficulty in distinguishing latently infected cells
Alfredo J Hernandez et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(1), 415-425 (2019-12-25)
Transposable elements make up half of the mammalian genome. One of the most abundant is the short interspersed nuclear element (SINE). Among their million copies, B2 accounts for ∼350,000 in the mouse genome and has garnered special interest because of
Yojet Sharma et al.
Wellcome open research, 5, 16-16 (2020-03-21)
The development of the human brain involves multiple cellular processes including cell division, migration, and dendritic growth. These processes are triggered by developmental cues and lead to interactions of neurons and glial cells to derive the final complex organization of
Aline M Thomas et al.
Journal of cellular physiology, 235(6), 5120-5129 (2019-11-02)
Dendritic cells are key players in regulating immunity. These cells both activate and inhibit the immune response depending on their cellular environment. Their response to hyperglycemia, a condition common amongst diabetics wherein glucose is abnormally elevated, remains to be elucidated.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service