Skip to Content
Merck
All Photos(1)

Key Documents

407318

Sigma-Aldrich

β-Interferon, Human, Recombinant, CHO Cells

Synonym(s):

Interferon α-2 precursor, IFN-β-1a, rHuIFN-β-1a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥98% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human interferon-β (IFN-β) expressed in CHO cells as a single glycosylated polypeptide containing 166 amino acids. Purified by chromatographic methods. IFN-β is a type I cytokine produced in response to exposure to viruses or double-stranded RNAs. It is also induced by other cytokines, such as TNF and IL-1. It is involved in the regulation of non-specific humoral immune responses and immune responses to viral infections, and thus exhibits anti-viral, anti-proliferative, and immunomodulating properties. The biological activity of IFN-β, unlike IFN-α, is strictly species-specific and will not be active across species.

Biochem/physiol Actions

Reported as IU/mg on a lot-specific basis using a viral resistance assay

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 62 mg/ml mannitol, 9.1 mg/ml human albumin, 0.9 mg/ml acetate, pH 7.0-7.2.

Reconstitution

Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 7 days at 4°C or for up to 4 months at -20°C. Avoid freeze/thaw cycles of solutions.
Reconstitute with sterile water to ≥100 µg/ml, containing 0.1% HSA or BSA. Further dilute with aqueous buffer just prior to use.

Other Notes

Goebel, M.U., et al. 2005. Physiol. Behav.84, 807.
Graber, J., et al. 2005. J. Neuroimmunol.161,169.
Mallada-Frechin, J., et al. 2005. Rev. Neurol.40, 482.
Patten, S.B., et al. 2005. Mult. Scler.11, 175.
Pohl, D., et al. 2005. Neurology64, 888.
Schwid, S.R., et al. 2005. Arch. Neurol.62, 785.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Taniguchi et al.
Journal of virology, 96(7), e0004922-e0004922 (2022-03-24)
Heartland bandavirus (HRTV), which is an emerging tick-borne virus first identified in Missouri in 2009, causes fever, fatigue, decreased appetite, headache, nausea, diarrhea, and muscle or joint pain in humans. HRTV is genetically close to Dabie bandavirus, which is the
Matthew Charman et al.
Frontiers in cellular and infection microbiology, 11, 689707-689707 (2021-10-09)
The induction of antiviral effector proteins as part of a homeostatically controlled innate immune response to infection plays a critical role in limiting the propagation and transmission of respiratory pathogens. However, the prolonged induction of this immune response can lead

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service