Skip to Content
Merck
All Photos(1)

Key Documents

343922

Sigma-Aldrich

5-Fluorouracil

≥98% (HPLC), solid, thymidylate synthase inhibitor, Calbiochem®

Synonym(s):

5-Fluorouracil, 5-FU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3FN2O2
CAS Number:
Molecular Weight:
130.08
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

5-Fluorouracil, Potent antitumor agent.

description

Merck USA index - 14, 4181

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

hot water: 1 mg/mL
methanol: 1 mg/mL
DMF: 5 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1[c]([nH][c]([nH]c1)=O)=O

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

GHASVSINZRGABV-UHFFFAOYSA-N

General description

Potent antitumor agent that inhibits aminoisobutyrate-pyruvate aminotransferase. Induces apoptosis of human colon cancer cell lines.
Potent antitumor agent. Inhibits aminoisobutyrate-pyruvate aminotransferase. Induces apoptosis in human colon cancer cell lines.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Aminoisobutyrate-pyruvate aminotransferase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Pressacco, J., et al. 1996. Br. J. Cancer72, 939.
Shchepotin, I.B., et al. 1994. Anticancer Res.14, 1027.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Germain et al.
Oncology, 102(6), 515-524 (2023-11-27)
Cancers in general, and specifically lung cancer, continue to have low patient survival rates when the patient is at an advanced stage when diagnosed. It appears that the local environment, especially fibroblasts and their signaling molecules, tends to induce metastasis
Chunfeng Liu et al.
Cell death and differentiation, 30(7), 1771-1785 (2023-05-21)
Curcumin, a natural phytochemical isolated from tumeric roots, represents a candidate for prevention and therapy of colorectal cancer/CRC. However, the exact mechanism of action and the downstream mediators of curcumin's tumor suppressive effects have remained largely unknown. Here we used

Articles

Discover critical characteristics to consider when working with enzyme inhibitors, such as cell permeability, the prozone effect, and Lipinski’s rule of 5.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service