Skip to Content
Merck
All Photos(1)

Documents

PHR1227

Supelco

Fluorouracil

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

5-Fluorouracil, 2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3FN2O2
CAS Number:
Molecular Weight:
130.08
Beilstein:
127172
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 995
traceable to Ph. Eur. F0250000
traceable to USP 12790000

API family

fluorouracil

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

282-286 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

FC1=CNC(=O)NC1=O

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

GHASVSINZRGABV-UHFFFAOYSA-N

Gene Information

human ... TYMS(7298)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Fluorouracil, an analog of the pyrimidines, belongs to the class of antimetabolite drugs. It is generally utilized for the remediation of tumors such as breast adenocarcinoma, gastrointestinal tract, ovary, head and neck.

Application

Fluorouracil may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by different analytical techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3468 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymeric nanoparticles for oral delivery of 5-fluorouracil: Formulation optimization, cytotoxicity assay and pre-clinical pharmacokinetics study
Mattos ACD, et al.
European Journal of Pharmaceutical Sciences, 84(11), 83-91 (2016)
Analysis of pharmaceutical creams: a useful approach based on solid-phase extraction (SPE) and UV spectrophotometry
Bonazzi D, et al.
Journal of Pharmaceutical and Biomedical Analysis, 13(11), 1321-1329 (1995)
Analysis of flucytosine dosage forms by derivative UV spectroscopy and liquid chromatography.
Cavrini V, et al.
Journal of Pharmaceutical and Biomedical Analysis, 9(5), 401-407 (1991)
Tania Diaz et al.
Journal of surgical oncology, 109(7), 676-683 (2014-02-11)
Surgery is the standard treatment for colorectal cancer (CRC), and adjuvant chemotherapy has been shown to be effective in stage III but less so in stage II. We have analyzed the expression of the miR-200 family in tissue samples from
André Bp van Kuilenburg et al.
Pharmacogenomics, 14(7), 799-811 (2013-05-09)
5-fluorouracil (5-FU) remains the cornerstone of all currently applied regimens for the treatment of patients with cancers of the gastrointestinal tract, breast, and head and neck. Unfortunately, a large variation in the clearance of 5-FU has been observed between patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service