260964
DNA-PK Inhibitor V
The DNA-PK Inhibitor V, also referenced under CAS 404009-46-7, controls the biological activity of DNA-PK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
DNA-PK Inhibitor V, AMA37, Aryl Morpholine Analog 37, 1-(2-Hydroxy-4-morpholin-4-yl-phenyl)-phenyl-methanone
About This Item
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
yellow
solubility
DMSO: 5 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
N3(CCOCC3)c1cc(c(cc1)C(=O)c2ccccc2)O
InChI
1S/C17H17NO3/c19-16-12-14(18-8-10-21-11-9-18)6-7-15(16)17(20)13-4-2-1-3-5-13/h1-7,12,19H,8-11H2
InChI key
FALILNHGILFDLC-UHFFFAOYSA-N
General description
Biochem/physiol Actions
DNA-PK
Packaging
Warning
Reconstitution
Other Notes
Willmore, E., et al. 2004. Blood,103, 4659.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service