Skip to Content
Merck
All Photos(1)

Key Documents

239764

Sigma-Aldrich

Cycloheximide, High Purity

Antifungal antibiotic that inhibits protein synthesis in eukaryotes but not in prokaryotes.

Synonym(s):

Cycloheximide, High Purity

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

potency

3.4 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

chloroform: soluble
ethanol: soluble
methanol: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C15H23NO4/c1-8-3-9(2)15(20)11(4-8)12(17)5-10-6-13(18)16-14(19)7-10/h8-12,17H,3-7H2,1-2H3,(H,16,18,19)/t8-,9-,11-,12+/m0/s1

InChI key

YPHMISFOHDHNIV-FSZOTQKASA-N

General description

Antifungal antibiotic that does not inhibit several species of pathogenic bacteria at 100 µg/ml. Inhibits protein synthesis in eukaryotes but not in prokaryotes. Interacts directly with the translocase enzyme, interfering with the translocation step. Inhibits cell-free protein synthesis in eukaryotes. Competitively inhibits hFkBP12 (Ki = 3.4 µM). Triggers apoptosis in HL-60 cells, T-cell hybridomas, Burkitt′s lymphoma cells, and a variety of other cell types, including rodent macrophages. Inhibits DNA cleavage in rat thymocytes treated with thapsigargin, methylprednisolone, and ionomycin. Rapidly destroyed in alkaline solutions.
Antifungal antibiotic that inhibits protein synthesis in eukaryotes but not in prokaryotes. Interacts directly with the translocase enzyme, interfering with the translocation step. Inhibits cell-free protein synthesis in eukaryotes. Competitively inhibits hFKBP12 (Ki = 3.4 µM). Triggers apoptosis in HL-60 cells, T cell hybridomas, Burkitt’s lymphoma cells, and a variety of other cell types including rodent macrophages. However, it inhibits DNA cleavage in rat thymocytes treated with Thapsigargin (Cat. No. 586005), methylprednisolone, and Ionomycin (Cat. Nos. 407950 and 407952). Rapidly destroyed in alkaline solutions. Also available as a 300 mM solution in DMSO (Cat. No. 508739).

Biochem/physiol Actions

Primary Target
hFKBP12

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 6 weeks at 4°C, pH 3-5.

Other Notes

Christner, C., et al. 1999. J. Med. Chem. 42, 3615.
Lu, Q., et al. 1996. Arch. Biochem. Biophys.334, 175.
Chow, S.C., et al. 1995. Exp. Cell Res.216, 149.
Cotter, T.G., et al. 1992. Anticancer Res.12, 773.
Takano, Y.S., et al. 1991. J. Pathol.163, 329.
Waring, P. 1990. J. Biol. Chem. 265, 14476.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Muta. 2 - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dirk Brehmer et al.
Molecular cancer therapeutics, 20(12), 2317-2328 (2021-09-30)
The protein arginine methyltransferase 5 (PRMT5) methylates a variety of proteins involved in splicing, multiple signal transduction pathways, epigenetic control of gene expression, and mechanisms leading to protein expression required for cellular proliferation. Dysregulation of PRMT5 is associated with clinical
Ashley N Stewart et al.
Biochemistry, 58(6), 727-741 (2018-12-20)
C1q/TNF-related protein 12 (CTRP12) is a secreted regulator of glucose and lipid metabolism. It circulates in plasma as a full-length protein or as a cleaved isoform generated by furin/PCSK3 cleavage. These isoforms preferentially activate different signaling pathways, and their ratio
Yuan-Lin Kang et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(34), 20803-20813 (2020-08-09)
Virus entry is a multistep process. It initiates when the virus attaches to the host cell and ends when the viral contents reach the cytosol. Genetically unrelated viruses can subvert analogous subcellular mechanisms and use similar trafficking pathways for successful
Cai Han et al.
STAR protocols, 3(1), 101037-101037 (2022-01-04)
Micropeptides are emerging as important regulators of various cellular processes. Long non-coding RNAs (lncRNAs) serve as a source of micropeptide-encoding small reading frames. The techniques to detect micropeptides or translating lncRNAs, such as mass spectrometry and ribosome profiling, are sophisticated
Songsong Dan et al.
Scientific reports, 11(1), 10285-10285 (2021-05-15)
Reversible lysine methylation is essential for regulating histones and emerges to critically regulate non-histone proteins as well. Here we show that the master transcription factor OCT4 in pluripotent stem cells (PSCs) was methylated at multiple lysine residues. LSD1 that is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service