Skip to Content
Merck
All Photos(1)

Key Documents

1.93502

Supelco

Sodium hydroxide

pellets, EMPLURA®

Synonym(s):

Sodium hydroxide, ‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
Pricing and availability is not currently available.

grade

laboratory grade

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

product line

EMPLURA®

Assay

99%

form

pellets
solid

pH

>14 (20 °C, 100  g/L)

bp

1,390 °C/1,013 hPa

mp

318 °C (lit.)

solubility

water: 1,090 g/L at 20 °C

density

2.13 g/cm3 at 20 °C

storage temp.

20-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93502.0521: 500G, Minimum order quantity: 20

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service