Skip to Content
Merck
All Photos(2)

Key Documents

189410

Sigma-Aldrich

Aurora Kinase Inhibitor VI, ZM447439

The Aurora Kinase Inhibitor VI, ZM447439, also referenced under CAS 331771-20-1, controls the biological activity of Aurora Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Aurora Kinase Inhibitor VI, ZM447439, 4-(4-(N-Benzoylamino)anilino)-6-methoxy-7-(3-(1-morpholino)propoxy)quinazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H31N5O4
CAS Number:
Molecular Weight:
513.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CC=C1)NC(C=C2)=CC=C2NC3=NC=NC4=C3C=C(OC)C(OCCCN5CCOCC5)=C4

InChI

1S/C29H31N5O4/c1-36-26-18-24-25(19-27(26)38-15-5-12-34-13-16-37-17-14-34)30-20-31-28(24)32-22-8-10-23(11-9-22)33-29(35)21-6-3-2-4-7-21/h2-4,6-11,18-20H,5,12-17H2,1H3,(H,33,35)(H,30,31,32)

InChI key

OGNYUTNQZVRGMN-UHFFFAOYSA-N

General description

A cell-permeable quinazoline compound that targets the ATP binding pocket and an adjacent cleft and acts as a potent and reversible inhibitor of Aurora A and Aurora B (IC50 = 110 and 130 nM, respectively) with moderate to excellent selectivity among closely related 16-kinase panel (IC50 = 0.88, 1.03 and 1.79 µM for Lck, Src and MEK1, respectively, and IC50 >10 µM for Cdk1/2/4, Plk1, Chk1). Blocks phosphorylation of Histone H3-Ser10 in Xenopus cycling egg extracts at 20 µM, and reduces mitotic spindle assembly. Shown to repress kinetochore localization of BubR1, Cenp-E, and Mad2, and arrest the growth of proliferating tumor cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Gadea, B.B. and Ruderman, J.V., 2005. Mol. Biol. Cell16, 1305.
Ditchfield, C., et al. 2003. J. Cell Biol.161, 267.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Onur Sen et al.
Developmental cell, 56(22), 3082-3099 (2021-11-11)
Chromosome mis-segregation during mitosis leads to aneuploidy, which is a hallmark of cancer and linked to cancer genome evolution. Errors can manifest as "lagging chromosomes" in anaphase, although their mechanistic origins and likelihood of correction are incompletely understood. Here, we
Federico Gulluni et al.
Science (New York, N.Y.), 374(6573), eabk0410-eabk0410 (2021-12-10)
Cytokinetic membrane abscission is a spatially and temporally regulated process that requires ESCRT (endosomal sorting complexes required for transport)–dependent control of membrane remodeling at the midbody, a subcellular organelle that defines the cleavage site. Alteration of ESCRT function can lead

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service