Skip to Content
Merck
All Photos(1)

Key Documents

SML3038

Sigma-Aldrich

JB170

≥98% (HPLC)

Synonym(s):

4-((9-chloro-7-(2-fluoro-6-methoxyphenyl)-5H-benzo[c]pyrimido[4,5-e]azepin-2-yl)amino)-N-(2-(2-(2-(2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamido)ethoxy)ethoxy)ethyl)-2-methoxybenzamide, Thalidomide-PEG2-Alisertib

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$158.00
25 MG
$637.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$158.00
25 MG
$637.00

About This Item

Empirical Formula (Hill Notation):
C48H44ClFN8O11
CAS Number:
Molecular Weight:
963.36
UNSPSC Code:
12352107
NACRES:
NA.77

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C1N(C2CCC(NC2=O)=O)C(C3=C1C(OCC(NCCOCCOCCNC(C4=CC=C(NC5=NC(C(C=CC(Cl)=C6)=C6C(C7=C(OC)C=CC=C7F)=NC8)=C8C=N5)C=C4OC)=O)=O)=CC=C3)=O

InChI key

GYKNPXCQINZRLL-UHFFFAOYSA-N

Biochem/physiol Actions

JB170 is a highly specific AURORA-A degrader (PROTAC) comprising aurora kinase inhibitor alisertib connected to the CEREBLON-binding molecule thalidomide. JB170 induces rapid and durable degradation of AURORA-A in cancer cells leading to strong S-phase arrest and apoptosis.
highly specific AURORA-A degrader (PROTAC) that induces rapid and durable degradation of AURORA-A in cancer cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bikash Adhikari et al.
Nature chemical biology, 16(11), 1179-1188 (2020-09-30)
The mitotic kinase AURORA-A is essential for cell cycle progression and is considered a priority cancer target. Although the catalytic activity of AURORA-A is essential for its mitotic function, recent reports indicate an additional non-catalytic function, which is difficult to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service