Skip to Content
Merck
All Photos(1)

Key Documents

1.88062

Supelco

Anolyte K for Coulometry

Coulometric Karl Fischer reagent for aldehydes & ketones, Aquastar®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Anolyte K for Coulometry, coulometric Karl Fischer reagent for aldehydes & ketones AQUASTAR®

product line

Aquastar®

form

liquid

reaction suitability

reaction type: coulometric

technique(s)

Karl-Fischer titration: suitable
titration: suitable

suitability

passes test for application

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Corr. 1C - STOT RE 1 Oral - STOT RE 2 - STOT SE 1

Target Organs

Immune system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. E' adatto per il KF coulometrico senza diaframma?

    1 answer
    1. The Anolyte K and Catholyte K reagents are suitable for use with cells with a diaphragm, only.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service