Skip to Content
Merck
All Photos(1)

Documents

1.73501

Supelco

Absorption tube for arsenic

joint: ST/NS 29

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

Joint

joint: ST/NS29

technique(s)

photometry: suitable

compatibility

for use with Spectroquant®

storage temp.

2-30°C

General description

This absorption tube is a required accessory when performing the Spectroquant® Arsenic test (1.01747). The tube is filled with one of the reagents of the kit and attached to the Erlenmeyer flask during the procedure. Afterwards the arsenic content of the sample can be determined from the solution in the absorbance tube using a Spectroquant® spectrophotometer. With ground joint NS 29 Spectroquant®.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service