Skip to Content
Merck
All Photos(6)

Documents

MMSMCTS48

Millipore

Milliflex Oasis® Culture Media Plate

Tryptic soy media plates for aerobic and anaerobic microflora

Synonym(s):

Green coded, ready-to-use agar plate for use with Milliflex Oasis® system, MILLIFLEX OASIS® Tryptic Soy Media Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

material

green polystyrene plate

Quality Level

Agency

complies with EP
complies with JP
complies with USP

sterility

sterile

form

plate (Pre-poured agar)

shelf life

12 mo.

packaging

pk of 48 tests

manufacturer/tradename

Milliflex Oasis®

pH

7.3±0.2 (25 °C)

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

compatibility

for use with MILLIFLEX OASIS® system

storage temp.

2-25°C

suitability

heterotrophic bacteria

Looking for similar products? Visit Product Comparison Guide

General description

MILLIFLEX OASIS® tryptic soy media plates are ready-to-use agar-based media for convenient, rapid and reliable testing. These unique green colour coded plates offer the control of critical handling steps all along the workflow in conjugation with the MILLIFLEX OASIS® system. They are manufactured and QC tested according to pharmaceutical industry requirements. The plates can be stacked and locked to ensure safe transportation and avoid accidental opening. The empty funnel tray can also be used as an incubation rack.

MILLIFLEX® TRACE features ensures total traceability through unique 2D barcodes identification and detachable labels. If left unopened, the plates remain sealed unlike standard petri plates which reduces condensation and dehydration for an extended storage time owing to its long shelf life.

Application

MILLIFLEX OASIS® tryptic soy media plates are used for the recovery of a broad range of heterotrophic microorganisms such as common aerobic and facultative anaerobic bacteria found in various types of water, Beer and Bottled Water. They are widely used in pharmaceutical and cosmetic industry.

Physical form

48 ready-to-use pre-poured agar plates

Analysis Note

The Colonies appear clear to creamy white, some may produce pigment

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex Oasis® system monitors bioburden in various samples, increasing lab productivity.

Milliflex® Rapid System 2.0 accelerates microbial detection in quality control labs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service