Skip to Content
Merck
All Photos(1)

Key Documents

1.52358

Supelco

Chromolith® WP 300 Protein A (monolithic) HPLC Columns

L × I.D. 25 mm × 2 mm HPLC Column

Synonym(s):

Chromolith® WP 300 Protein A 25-2 HPLC column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Chromolith® WP 300 Protein A, L × I.D. 25 mm × 2 mm HPLC column

Quality Level

feature

endcapped: no

parameter

2-45 °C temp. range
200 bar pressure

technique(s)

HPLC: suitable

column L × I.D.

25 mm × 2 mm

matrix

monolithic silica

matrix active group

Protein A bonding phase

pore size

2 μm macroporosity
30 nm mesoporosity

operating pH

1.5-7.5

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Affinity chromatography is a selective technique that takes advantage of specific molecular interactions, for example, antigen and antibody. The Chromolith® WP 300 Protein A HPLC column is designed to monitor monoclonal antibody titer and yield determination from cell-culture supernatants. This analytical scale procedure helps to optimize the titer of monoclonal antibody for the optimal time for harvest of the monoclonal antibody products.

Chromolith® WP 300 Protein A, 25-2 columns are made of monolithic silica with a characteristic bimodal pore structure.

Key Benefits:
  • Rapid and accurate titer determination across a wide range of injection volumes
  • Consistent binding efficiencies even at high flow rates
  • Expanded column lifetimes


Learn more:
Flyer: Rapid and Robust Antibody Quantitation - Chromolith® WP 300 Protein A HPLC columns

Brochure: Race through your Separations on Any System. Chromolith® Monolithic Silica HPLC Columns

All Supelco HPLC Columns, including monolithic Chromolith® columns, are fully compatible to all HPLC and UHPLC Instruments.
Supelco HPLC Products: The Best Choice for Any LC Instrument

Legal Information

Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Method development for titer determination of three different monoclonal antibodies - cetuximab, trastuzumab, and universal antibody standard using a Chromolith® WP 300 Protein A column.

Related Content

Analytical Chromolith® HPLC columns for Bio applications, 300A mesopores: RP-18, RP-8, RP-4, Prot-A, Epoxy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service