Skip to Content
Merck
All Photos(1)

Documents

55215-U

Supelco

Supelco® ColumnSaver Precolumn Filter

size 2.0 μm, pkg of 10 ×  ea

Synonym(s):

ColumnSaver precolumn guard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

packaging

pkg of 10 ×  ea

size

2.0 μm

Looking for similar products? Visit Product Comparison Guide

General description

  • Economical protection for your guard or analytical HPLC column
  • No wrenches or tools required to install, fingertight to 5,000 psi
  • Lower dead volume than conventional filters with holders
  • Universal connection is compatible with all manufacturers fittings
·
The new Supelco ColumnSaver offers all of the protection of conventional precolumn filters at much less the cost. With its convenient direct connect design, changeover time is measured in seconds, and requires no wrenches or tools to install.

The PEEK filter body contains a HiFlo filter element and is designed for maximum filtration of particulate matter with mimimal dead volume or backpressure. As soon as an increase in backpressure is observed, simply remove and dispose of the Supelco ColumnSaver and install a new one. The direct connect design is compatible with all 1/16 in., 10-32 internal fitting ports regardless of the manufacturer.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service