Skip to Content
Merck
All Photos(1)

Key Documents

1.50187

Supelco

LiChroCART 25-4 LiChrospher® RP-18 ADS

for use with LICHROCART 25-4, LiChrospher® RP-18 ADS, MANU-CART

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
Pricing and availability is not currently available.

Quality Level

technique(s)

solid phase extraction (SPE): suitable

compatibility

for use with LICHROCART 25-4, LiChrospher RP-18 ADS, MANU-CART

storage temp.

no temp limit

General description

HPLC cartridge set, contains 1 LiChroCART 25-4 LiChrospher RP-18 ADS 1 manu-CART
LiChrospher® ADS allows the fully automated, LC integrated sample preparation of hydrophobic, low molecular weight analytes from untreated samples like serum, milk, fermentation broth of food homogenates. The enrichment is based on two chromatographic processes: reversed phase and size exclusion chromatography since LiChrospher ADS sorbents are spherical silica gel particles (25µm) with DIOL modification on the exterior surface and alkyl modifications on the surface of the interior pores (ID: 6nm). Depending on the hydrophobicity of the analyte three types of precolumns are available: C-4, C-8 and C-18.

Analysis Note

visual control (for delivery): passes test
judgement (performance): passes test

Legal Information

LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany
MANU-CART is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service