Skip to Content
Merck
All Photos(1)

Key Documents

14-789-M

Sigma-Aldrich

PI 3-Kinase (p110δ/p85α) murine, 10 µg

Complex of N-terminal 6His-tagged recombinant mouse p110δ full length & untagged, recombinant, mouse p85α full length.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

mouse

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 124 kDa (p110δ)
Mw 84 kDa (p85α)

manufacturer/tradename

Upstate®

NCBI accession no.

General description

Complex of N-terminal 6His-tagged recombinant mouse p110δ full length and untagged, recombinant, mouse p85α full length.

Packaging

Also available in 250μg size --call for pricing and availability and reference catalog number 14-789M when ordering the 250μg size.

Physical form

50 mM Tris/HCl pH7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 0.2 mM PMSF, 1 mM benzamidine, 0.1% 2-mercaptoethanol. Frozen solution.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service