Skip to Content
Merck
All Photos(2)

Key Documents

1.46784

Millipore

Tryptic Soy Agar - Ready-to-use Contact Plates

IsoBag®, Lecithin, Tween®, ICR plate with non-lockable lid, plate diam. 55 mm, DPTE Beta Bag with 190 mm alpha-port, sterile; irradiated, single packed of 10 × 10 plates, suitable for surface monitoring

Synonym(s):

IsoBag® Tryptic Soy Agar Contact, Isolator contact plate, Soybean Casein Digest Agar, TSA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106231
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

material

polystyrene plate

Agency

EP
USP

sterility

sterile; irradiated

form

plate

feature

rapid transfer port
ready-to-use
DPTE Beta Bag with 190 mm alpha-port

composition

Lecithin
Tween®

packaging

single packed of 10 × 10 plates

manufacturer/tradename

IsoBag®

storage condition

protect from light (Keep tightly closed and dry)

plate diam.

55 mm

color

light yellow

application(s)

pharmaceutical
surface monitoring

compatibility

stand alone

suitability

bacteria
molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

The IsoBag® Tryptic Soy Agar Contact is designed for quick and convenient transfer of culture media for surface monitoring into isolators or RABS systems. The IsoBag® consists of a polyurethane DPTE Beta Bag compatible with 190 mm Alpha Ports. The bag contains 10 stacks (single wrapped) of contact plates and sterile Polyethylene zip lock bags to mitigate the risk of a cross contamination during transport of the plates. Tryptic Soy Agar is supplemented with Lecithin, polysorbate (Tween®) 80 and pyruvate to neutralize residues of disinfectants. The IsoBag® is designed to reduce the decontamination cycles in production isolators.

Application

The IsoBag® Tryptic Soy Agar Contact is a sterile polyurethane DPTE Beta Bag designed for quick and convenient transfer of culture media used for surface monitoring in isolators or RABS.

Legal Information

ISOBAG is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

IsoBag® ease the environmental monitoring workflow in aseptic production isolator

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service