1.46685
Tryptic Soy Agar - Ready-to-use Settle Plates
ICR with lockable lid, plate volume × diam. 30 mL × 90 mm, sterile; γ-irradiated, pkg of 10 plates Triple packed, suitable for air monitoring
Synonym(s):
CASO Agar, Casein Soybean Digest Agar, TSA, Tryptic Soy Agar
About This Item
Recommended Products
material
polystyrene plate
Quality Level
Agency
EP
USP
sterility
sterile; γ-irradiated
form
plate
feature
ready-to-use
packaging
pkg of 10 plates Triple packed
box of 120 plates 1466850120
box of 20 plates
storage condition
protect from light (Tightly closed. Dry)
plate volume × diam.
30 mL × 90 mm
color
light yellow
pH
7.3 ( in H2O)
application(s)
air monitoring
pharmaceutical
compatibility
for use with MAS-100®
storage temp.
2-25°C
suitability
bacteria
fungi
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+
Related Content
Settle plates for isolators and critical cleanrooms
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service