Skip to Content
Merck
All Photos(1)

Key Documents

14-1109

Sigma-Aldrich

Human Recombinant Histone H2A.Z

Human recombinant histone H2A.Z expressed in E Coli & purified by Fast Protein Liquid Chromatography (FPLC) & verified mass spectrometry.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

Escherichia coli

Quality Level

recombinant

expressed in E. coli

Assay

>98% (SDS-PAGE)

concentration

1.0 μg/μL

technique(s)

activity assay: suitable

shipped in

dry ice

General description

Full-length human recombinant H2A.Z expressed in E. Coli and purified by Fast Protein Liquid Chromatography (FPLC ).
The histone variant H2A.Z is a evolutionarily conserved histone variant with a calculated molecular weight of13.5 kDa. This variant replaces conventional histone H2A in a subset of nucleosomes. The nucleosome is the basic unit of chromatin and consists of 147 base pairs of DNA wrapped around an octamer of core histones H2A, H2B, H3 and H4. The histone variant H2AZ has emerged as a key regulator of chromatin function and plays an essential role during mammalian development. H2A.Z is involved in transcriptional regulation, antisilencing, silencing, and genome stability.

Application

Suitable for in vitro chromatin assembly and enzyme activity assays

Physical form

Recombinant protein in a buffer containing 20mM Tris-Cl pH 7.6, 1mM EDTA, 0.15 M NaCl, 10% glycerol, 0.5mM PMSF and 1mM DTT.

Storage and Stability

Stable at -80ºC for 1 year from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service