Skip to Content
Merck
All Photos(1)

Documents

1.17337

Millipore

GranuCult® plus m-TGE (modified Tryptone Glucose Meat extract) broth

nonselective for, suitable for microbiological culture

Synonym(s):

TGE Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

EP 2.6.1
JP 4.06
USP 71

Sterilization Compatibility

autoclavable compatible

form

powder

packaging

pkg of 500 g

technique(s)

microbiological culture: suitable

application(s)

food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

nonselective for

General description

GranuCult® plus m-TGE (modified Tryptone Glucose Meat extract) broth also known as TGE broth is a non-selective microbial media. The media is compatible with samples without inhibitors.
Tryptone and meat extract present in the formulation forms the nitrogen and carbon source along with supplying essential nutrients. And glucose in the medium provides energy for microbial growth.

Application

GranuCult® plus m-TGE (modified Tryptone Glucose Meat extract) broth is used to detect total heterotrophic microorganisms in water, dairy products, and other products like beer, bottled water, cider, soft drinks, and wine samples by membrane filtration technique.
It is also used for the detection of thermophilic bacteria in milk and dairy products.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service