Skip to Content
Merck
All Photos(1)

Key Documents

Z699004

M-TGE Broth

pack of 50 × 2 mL

Synonym(s):

M-TGE Culture Broth, TGE Broth

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$264.00

$264.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
50 EA
$264.00

About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

$264.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pack of 50 × 2 mL

manufacturer/tradename

Maine Mfg 10496102

application(s)

food and beverages

storage temp.

2-8°C

General description

All bacteria develop on TGE media and produce a range of different colored and sized colonies.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service