Skip to Content
Merck
All Photos(1)

Key Documents

1.00683

Supelco

Ammonium Test, photometric

photometric, 2.0-150 mg/L (NH4-N), 2.6-193 mg/L (NH4+), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Ammonium Test, photometric, 2.0-150 mg/L (NH4-N), 2.6-193 mg/L (NH4+), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests

specific analyte(s)

ammonium

measuring range

2.0-150 mg/L (NH4-N)
2.4-193 mg/L (NH3)
2.6-193 mg/L (NH4+)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Indophenol blue)

storage temp.

15-25°C

General description

Method: Photometric, 2.0 - 150 mg/L (NH4-N), 2.6 - 193 mg/L (NH4+), Spectroquant®
The method is analogous to EPA 350.1, APHA 4500-NH3 F, ISO 7150-1, and DIN 38406-5.
This Spectroquant® Ammonium Reagent Test allows the cyanide-free quantification of ammonium in wastewater, groundwater, surface water, and seawater. Typically, ammonium enters water sources as run-off from fertilized agricultural areas and enhanced levels are important indicators for fecal pollution.
Further fields of applications are the analysis of foods & beverages and of environmental and agricultural samples (e.g. soil).
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Ammonium is the naturally preferred source of nitrogen for plant growth and there is an increasing global demand for organic food products.

Step-by-step method describing the photometric determination of ammonium ions in effluents using Spectroquant® test kits and spectrophotometer.

Photometric determination of ammonium ions in organic fertilizers by indophenol blue method using Spectroquant® test kits and spectrophotometer.

Photometric determination of ammonium ions in mineral fertilizer samples by indophenol blue method using Spectroquant® test kits and spectrophotometer.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service