Skip to Content
Merck
All Photos(1)

Documents

1.10863

Millipore

Tetrathionate enrichment broth base acc. to MULLER-KAUFFMANN

for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.6 (37 °C, 82 g/L in H2O, after autoclaving)

solubility

82 g/L

bulk density

1100 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Salmonella spp.

Related Categories

Analysis Note

Appearance (clearness): turbid with sediment
Appearance (colour): green
Inoculum (Escherichia coli ATCC 25922 (WDCM 00013)): about 99 %
Inoculum (Salmonella typhimurium ATCC 14028 (WDCM 00031)): about 1 %
Enrichment from a mixed inoculum after 6 hours (Escherichia coli ATCC 25922 (WDCM 00013)):
Enrichment from a mixed inoculum after 6 hours (Salmonella typhimurium ATCC 14028 (WDCM 00031)):
Enrichment from a mixed inoculum after 24 hours (Escherichia coli ATCC 25922 (WDCM 00013)): ≤ 5 %
Enrichment from a mixed inoculum after 24 hours (Salmonella typhimurium ATCC 14028 (WDCM 00031)): ≥ 95 %
Incubation: 35 °C; aerobic; detection with BROLACIN agar.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service