Skip to Content
Merck
All Photos(1)

Key Documents

1.05450

Millipore

GranuCult® Nutrient agar

ISO 6579, ISO 10273, ISO 19250, ISO 21528, APHA, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

APHA
ISO 10273
ISO 19250
ISO 21528
ISO 6579

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.0 (37 °C, 20 g/L in H2O, after autoclaving)

solubility

20 g/L

bulk density

600 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

heterotrophic bacteria

General description

GranuCult® Nutrient agar is a universal culture media for cultivating less fastidious microorganisms. The peptone and beef extract provide sufficient nutrients for the growth of non-fastidious bacteria. Both are the sources of nitrogen, vitamins, carbon, and amino acids, while agar is the solidifying agent.

Application

GranuCult® Nutrient Agar is used for the cultivation of non-fastidious bacteria.

Analysis Note

Appearance (clearness): clear to slightly opalescent
Appearance (colour): yellowish-brown
pH-value (25 °C): 6.8 - 7.2
Solidification behaviour (2 hrs., 45 °C): liquid
Growth promotion test in accordance with the current version of DIN EN ISO 11133.
Inoculum on reference medium (Escherichia coli ATCC 8739 (WDCM 00012)):
Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013)):
Inoculum on reference medium (Salmonella typhimurium ATCC 14028 (WDCM 00031)):
Inoculum on reference medium (Salmonella enteritidis ATCC 13076 (WDCM 00030)):
Inoculum on reference medium (Yersinia enterocolitica ATCC 9610 (WDCM 00038)):
Inoculum on reference medium (Yersinia enterocolitica ATCC 23715 (WDCM 00160)):
Inoculum on reference medium (Staphylococcus aureus ATCC 25923 (WDCM 00034)):
Colony count (Escherichia coli ATCC 8739 (WDCM 00012)):
Colony count (Escherichia coli ATCC 25922 (WDCM 00013)):
Colony count (Salmonella typhimurium ATCC 14028 (WDCM 00031)):
Colony count (Salmonella enteritidis ATCC 13076 (WDCM 00030)):
Colony count (Yersinia enterocolitica ATCC 9610 (WDCM 00038)):
Colony count (Yersinia enterocolitica ATCC 23715 (WDCM 00160)):
Colony count (Staphylococcus aureus ATCC 25923 (WDCM 00034)):
Recovery on test medium (Escherichia coli ATCC 8739 (WDCM 00012)): ≥ 70 %
Recovery on test medium (Escherichia coli ATCC 25922 (WDCM 00013)): ≥ 70 %
Recovery on test medium (Salmonella typhimurium ATCC 14028 (WDCM 00031)): ≥ 70 %
Recovery on test medium (Salmonella enteritidis ATCC 13076 (WDCM 00030)): ≥ 70 %
Recovery on test medium (Yersinia enterocolitica ATCC 9610 (WDCM 00038)): ≥ 70 %
Recovery on test medium (Yersinia enterocolitica ATCC 23715 (WDCM 00160)): ≥ 70 %
Recovery (Staphylococcus aureus ATCC 25923 (WDCM00034)): ≥ 70 %
Incubation:
24 ± 2 at 37 ± 1 °C aerobic
Yersinia 24 ± 2 hours at 30 ± 1 °C aerobic
A recovery rate of 70 % is equivalent to a productivity value of 0.7.
The indicated colony counts result from the sum of a triple determination.
Reference media: Tryptic Soy Agar

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danielle Aparecida Duarte Nunes et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 55(6), 650-660 (2020-02-19)
Bioremediation is one of the existing techniques applied for treating oil-contaminated soil, which can be improved by the incorporation of low-cost nutritional materials. This study aimed to assess the addition of two low-cost plant residues, sugarcane bagasse (SCB) and leaf

Articles

Interview: Accurate microbiology QC methods for infant formula testing

Protocols

The International Organization for Standardization (ISO) published the revised EN ISO 21528-1 and EN ISO 21528-2 standards in 2017.

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service