Skip to Content
Merck
All Photos(1)

Key Documents

1.09905

Supelco

Bromide bromate solution

c(Br₂) = 0,05 mol/l (0.1 N), concentrate for 1000 mL titration solution, Titrisol®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Bromide bromate solution, for 1000 ml, c(Br₂) = 0,05 mol/l (0.1 N) Titrisol®

Quality Level

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Redox Reactions

concentration

0.05 M

technique(s)

titration: suitable

pH

12.8 (20 °C in H2O)

density

1.11 g/cm3 at 20 °C

storage temp.

15-25°C

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Ox. Liq. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hallo, ich suche ein Kaliumbromid-Kaliumbromat-Reagenz zur Quecksilberbestimmung mittels AAS gemäß EN ISO 12846:2012. Ist Ihr Produkt 1.09905 dafür geeignet? Könnten Sie mir bitte die genaue Konz von Kaliumbromid und Kaliumbromat in der Lösung mitteilen?

    1 answer
    1. Unfortunately, this product is a titrant and is not suitable for use in mercury vapor AAS. The reagent solution for this protocol should be prepared from low mercury potassium bromide and potassium bromate materials. Please see the links below to review components that are more suitable for this application:
      https://www.sigmaaldrich.com/US/en/product/mm/104912
      https://www.sigmaaldrich.com/US/en/product/mm/104905

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service