Skip to Content
Merck
All Photos(3)

Documents

1.07176

Supelco

n-Pentane

about 95%, EMPLURA®

Synonym(s):

Pentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-692-4
NACRES:
NA.21

product name

n-Pentane about 95%, EMPLURA®

vapor density

2.48 (vs air)

Quality Level

product line

EMPLURA®

Assay

≥95% (GC)

form

liquid

autoignition temp.

500 °F

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

~8.3 %

impurities

≤0.001% Free acid (as CH3COOH)

evapn. residue

≤0.005%

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

transition temp

flash point -48 °C (refers to pure substance)

solubility

0.4 g/L

density

0.626 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Oil sludge washing with surfactants and co-solvents: This study by Ramirez et al. explored the effectiveness of various solvents, including n-Pentane, in recovering oil from different types of oil sludges, focusing on environmental remediation technologies (Ramirez D, Shaw LJ, Collins CD, 2021).

  • Physicochemical properties of organic solvents in nanoparticle synthesis: Fromme et al. studied the impact of organic solvents, such as n-Pentane, on gas and hydrogen formation during laser synthesis of gold nanoparticles, highlighting its application in materials science (Fromme T, Tintrop LK, Reichenberger S, Schmidt TC, Barcikowski S, 2023).

  • Thermodynamic assessment in extraction processes: Al-Maari et al. assessed the efficiency of n-Pentane as a solvent in the extraction of vegetable oils using thermodynamic models, which is crucial for food and pharmaceutical industries (Al-Maari MA, Hizaddin HF, Salleh MZM, Hayyan A, 2024).

Analysis Note

Purity (GC): ≥ 95.0 %
Identity (IR): conforms
Free acid (as CH₃COOH): ≤ 0.001 %
Density (d 20 °C/ 4 °C): 0.626 - 0.628
Readily carbonizable substances: conforms
Evaporation residue: ≤ 0.005 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service