1.09731
Tetrahydrofuran
≥99.8% (GC), ACS reagent, reag. Ph. Eur., EMSURE®
Synonym(s):
THF, Tetramethylene oxide, Oxolane, Butylene oxide, Butylene oxide, Oxolane, Tetramethylene oxide
About This Item
Recommended Products
product name
Tetrahydrofuran, for analysis EMSURE® ACS,Reag. Ph Eur
grade
ACS reagent
Quality Level
Agency
reag. Ph. Eur.
vapor density
2.5 (vs air)
vapor pressure
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)
product line
EMSURE®
Assay
≥99.8% (GC)
form
liquid
autoignition temp.
610 °F
expl. lim.
1.8-11.8 %
impurities
≤0.00005% Al (Aluminium)
≤0.0002 meq/g Alkalinity
≤0.0003 meq/g Acidity
≤0.005% Peroxide (as H2O2)
≤0.03% Water
evapn. residue
≤0.0005%
color
clearAPHA: ≤10
refractive index
n20/D 1.407 (lit.)
pH
7-8 (20 °C, 200 g/L in H2O)
bp
65-67 °C (lit.)
mp
−108 °C (lit.)
transition temp
flash point -21.5 °C
density
0.889 g/mL at 25 °C (lit.)
cation traces
Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.000002%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%
storage temp.
2-30°C
SMILES string
C1CCOC1
InChI
1S/C4H8O/c1-2-4-5-3-1/h1-4H2
InChI key
WYURNTSHIVDZCO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Construction of Polycarbonates with Pendant Multifunctional Groups via a One-Step CO2/Diepoxide Copolymerization Approach.: This study employs tetrahydrofuran (THF) as a solvent in the synthesis of multifunctional polycarbonates through CO2 copolymerization, demonstrating its utility in creating advanced materials with potential applications in environmental sustainability and polymer chemistry (Wu et al., 2024).
Analysis Note
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Density (d 20 °C/20 °C): 0.885 - 0.895
Acidity: ≤ 0.0003 meq/g
Alkalinity: ≤ 0.0002 meq/g
Peroxides: conforms
Peroxide (as H₂O₂): ≤ 0.005 %
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.000002 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
Mg (Magnesium): ≤ 0.000002 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %
Evaporation residue: ≤ 0.0005 %
Water: ≤ 0.03 %
Stabilized with 2,6-Di-tert-butyl-4-methylphenol (BHT).
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system, Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-6.2 °F - closed cup
Flash Point(C)
-21.2 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service