Skip to Content
Merck
All Photos(2)

Key Documents

1.08110

Supelco

Tetrahydrofuran

Tetrahydrofuran
1 of 1 reviewers received a sample product or took part in a promotion

≥99.9% (GC), suitable for UV/Vis spectroscopy, Uvasol®

Synonym(s):

THF, Tetramethylene oxide, Oxolane, Butylene oxide, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-726-8
NACRES:
NA.03
Assay:
≥99.9% (GC)
technique(s):
UV/Vis spectroscopy: suitable
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Tetrahydrofuran, for spectroscopy Uvasol®

vapor density

2.5 (vs air)

Quality Level

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

610 °F

expl. lim.

1.8-11.8 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.01% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

215 nm, ≥30%
245 nm, ≥50%
265 nm, ≥80%
275 nm, ≥90%
310 nm, ≥98%

refractive index

n20/D 1.407 (lit.)

pH

7-8 (20 °C, 200 g/L in H2O)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

transition temp

flash point -20 °C

density

0.889 g/mL at 25 °C (lit.)

UV absorption

λ: 215 nm Amax: ≤0.52
λ: 245 nm Amax: ≤0.30
λ: 265 nm Amax: ≤0.10
λ: 275 nm Amax: ≤0.05
λ: 310 nm Amax: ≤0.01

storage temp.

2-30°C

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Purity (GC): ≥ 99.9 %
Evaporation residue: ≤ 0.0002 %
Water: ≤ 0.01 %
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Fluorescence (as quinine at 254 nm): ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm): ≤ 1.0 ppb
Transmission (at 215 nm): ≥ 30 %
Transmission (at 245 nm): ≥ 50 %
Transmission (at 265 nm): ≥ 80 %
Transmission (at 275 nm): ≥ 90 %
Transmission (from 310 nm): ≥ 98 %
Absorbance (at 215 nm): ≤ 0.52
Absorbance (at 245 nm): ≤ 0.30
Absorbance (at 265 nm): ≤ 0.10
Absorbance (at 275 nm): ≤ 0.05
Absorbance (from 310 nm): ≤ 0.01

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service