1.07414
Lead(II) hydroxide acetate
anhydrous for the analysis of sugar acc. to Horne EMSURE®
Synonym(s):
Lead(II) hydroxide acetate, Horne′s compound
About This Item
Quality Level
product line
EMSURE®
form
crystalline
concentration
≥33.0% (PbO)
impurities
≤0.02% Substances insoluble in dilute acetic acid
≤1% In water insoluble matter
loss
≤1% loss on drying, 105 °C
pH
7.2 (20 °C, 50 g/L in H2O)
mp
198-205 °C
density
3.58 g/cm3 at 22 °C
bulk density
2100 kg/m3
anion traces
chloride (Cl-): ≤0.003%
nitrate and nitrite (as NO3-): passes test
cation traces
Ca: ≤0.01%
Cu: ≤0.002%
Fe: ≤0.002%
K: ≤0.02%
Na: ≤0.05%
storage temp.
2-30°C
SMILES string
[Pb+2].[Pb].[O-]C(=O)C.[O-]C(=O)C.O.O
InChI
1S/2C2H4O2.2H2O.2Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);2*1H2;;/q;;;;;+2/p-2
InChI key
XAOPZCQFBUCRKR-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Analysis Note
In water insoluble matter: ≤ 1.0 %
Substances insoluble in dilute acetic acid: ≤ 0.02 %
Chloride (Cl): ≤ 0.003 %
Nitrate und Nitrite (as NO₃): passes test
Ca (Calcium): ≤ 0.01 %
Cu (Copper): ≤ 0.002 %
Fe (Iron): ≤ 0.002 %
K (Potassium): ≤ 0.02 %
Na (Sodium): ≤ 0.05 %
Loss on Drying (105 °C): ≤ 1.0 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service