Skip to Content
Merck
Lead(II) hydroxide acetate anhydrous for the analysis of sugar acc. to Horne EMSURE®

Lead(II) hydroxide acetate

Lead(II) hydroxide acetate anhydrous for the analysis of sugar acc. to Horne EMSURE®
Synonym(s):
Lead(II) hydroxide acetate
Linear Formula:
(CH3COO)2Pb · Pb(OH)2
CAS Number:
Molecular Weight:
566.50

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1