Skip to Content
Merck
All Photos(1)

Documents

1.07232

Millipore

BPLS agar

for the isolation of Salmonella

Synonym(s):

Brilliant-green phenol-red lactose sucrose agar, Brillant-green phenol-red lactose sucrose agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

6.8-7.0 (37 °C, 52 g/L in H2O, after autoclaving)

solubility

52 g/L

bulk density

600 kg/m3

application(s)

clinical
microbiology

storage temp.

15-25°C

suitability

Salmonella spp.

General description

for the isolation of Salmonella

Analysis Note

Appearance (clearness)clearAppearance (colour)dark redSolidification point32 - 36 °CGel strength (Gelomat assay)≥ 50 gpH-value (25 °C)6.7 - 7.1Inoculum on reference medium (Salmonella typhimurium ATCC 14028 (WDCM 00031))10 - 1000Inoculum on reference medium (Salmonella choleraesuis ATCC 13312)10 - 1000Inoculum on reference medium (Salmonella enteritidis NCTC 5188 (WDCM 00119))10 - 1000Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013))10 - 1000Inoculum on reference medium (Proteus hauseri ATCC 13315)10 - 1000Inoculum (Staphylococcus aureus ATCC 25923 (WDCM 00034))10⁴Inoculum (Enterococcus faecalis ATCC33186)10⁴Inoculum (Bacillus subtilis ATCC 6633 (WDCM 00003))10⁴Colony count (Salmonella typhimurium ATCC 14028 (WDCM 00031))Colony count (Salmonella choleraesuis ATCC 13312)Colony count (Salmonella enteritidis NCTC 5188 (WDCM 00119))Colony count (Escherichia coli ATCC 25922 (WDCM 00013))Colony count (Proteus hauseri ATCC 13315)Recovery on test medium (Salmonella typhimurium ATCC 14028 (WDCM 00031))≥ 40%Recovery on test medium (Salmonella choleraesuis ATCC 13312)≥ 40%Recovery on test medium (Salmonella enteritidis NCTC 5188 (WDCM 00119))≥ 40%Recovery on test medium (Escherichia coli ATCC 25922 (WDCM 00013))no LimitRecovery on test medium (Proteus hauseri ATCC 13315)no limitGrowth (Bacillus subtilis ATCC 6633 (WDCM 00003))no limitGrowth (Enterococcus faecalis ATCC 33186)no limitGrowth (Staphylococcus aureus ATCC 25923 (WDCM 00034))no limitColony colour (Salmonella typhimurium ATCC 14028 (WDCM 00031))pinkColony colour (Salmonella choleraesuis ATCC 13312)pinkColony colour (Salmonella enteritidis NCTC 5188 (WDCM 00119))pinkColony colour (Escherichia coli ATCC 25922 (WDCM 00013))yellowColony colour (Proteus hauseri ATCC 13315)yellowIncubation: 24 hrs.; 35 °C; aerobic.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service