Skip to Content
Merck
All Photos(1)

Documents

1.06794

Supelco

3-Nitrophenol

pH indicator

Synonym(s):

3-Nitrophenol, m-Nitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
209-073-5
NACRES:
NA.21

product name

3-Nitrophenol, indicator

Quality Level

Assay

≥98% (DSC (differential scanning calorimetry))

form

solid

autoignition temp.

400 °C

potency

328 mg/kg LD50, oral (Rat)

pH

6.6-8.6 (20 °C in H2O)

bp

194 °C/93 hPa

mp

95-98 °C

transition temp

flash point >100 °C

solubility

insoluble 13.5 g/L

density

1.49 g/cm3 at 20 °C

bulk density

640 kg/m3

λmax

329-333 nm (buffer pH 6.6)
389-393 nm (buffer pH 11.0)

storage temp.

2-30°C

InChI

1S/C6H5NO3/c8-6-3-1-2-5(4-6)7(9)10/h1-4,8H

InChI key

RTZZCYNQPHTPPL-UHFFFAOYSA-N

Related Categories

Analysis Note

Purity (DSC (differential scanning calorimetry)): ≥ 98 %
Identity (UV/VIS-Spectrum): passes test
Melting point (DSC (differential scanning calorimetry)): 94 - 98 °C
Transition range: pH 6.6 - pH 8.6colourless - yellow
Absorption maximum λmax. (buffer pH 6.6): 329 - 333 nm
Absorption maximum λmax. (buffer pH 11.0): 389 - 393 nm
Spec. Absorptivity A 1%/1cm (λ1max; 0.05 g/l; buffer pH 6.6): 130 - 150
Spec. Absorptivity A 1%/1cm (λ2max; 0.05 g/l; buffer pH 11.0): 100 - 120

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service