Skip to Content
Merck
All Photos(1)

Documents

1.06672

Supelco

Sodium tungstate dihydrate

EMPLURA®

Synonym(s):

Sodium tungstate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O4W · 2H2O
CAS Number:
Molecular Weight:
329.85
MDL number:
UNSPSC Code:
12352302
EC Index Number:
236-743-4
NACRES:
NA.21

Quality Level

product line

EMPLURA®

Assay

≥98% (thermogravimetric)

form

crystals

potency

1190 mg/kg LD50, oral (Rat)

pH

8-9 (20 °C, 50 g/L in H2O)

mp

100 °C (Elimination of water of crystallisation)

solubility

730 g/L

density

3.23 g/cm3 at 20 °C

bulk density

1600 kg/m3

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.002%
N: ≤0.005%
excess alkali (as NaOH): ≤0.1%

storage temp.

15-25°C

InChI

1S/2Na.2H2O.4O.W/h;;2*1H2;;;;;/q2*+1;;;;;2*-1;

InChI key

WPZFLQRLSGVIAA-UHFFFAOYSA-N

Application


  • Synthesis and characterization of Anderson-Evans type polyoxometalates, antibacterial properties.: This research focuses on the synthesis and characterization of Anderson-Evans type polyoxometalates, evaluating their antibacterial properties. Sodium tungstate dihydrate is used in the synthesis process to form these polyoxometalates. (Avci Özbek, 2023).

  • Nitric oxide modulates folate-mediated one-carbon metabolism and mitochondrial energy levels of peaches during cold storage.: This study explores the role of nitric oxide in modulating folate-mediated one-carbon metabolism and mitochondrial energy levels in peaches during cold storage. Sodium tungstate dihydrate is used as an analytical reagent in the experiments. (Yang et al., 2023).

  • Facile Bottom-up Preparation of WS(2)-Based Water-Soluble Quantum Dots as Luminescent Probes for Hydrogen Peroxide and Glucose.: Details a novel method for preparing WS(2)-based water-soluble quantum dots used as luminescent probes for detecting hydrogen peroxide and glucose. Sodium tungstate dihydrate is used in the preparation of these quantum dots. (Hang et al., 2019).

Analysis Note

Assay (thermogravimetric): ≥ 98 %
Identity (x-ray diffraction): passes test
Free alkali (as NaOH): ≤ 0.1 %
Chloride (Cl): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.02 %
Total nitrogen (N): ≤ 0.005 %
Fe (Iron): ≤ 0.002 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service