Skip to Content
Merck
All Photos(2)

Key Documents

1.02382

Supelco

Calcium chloride dihydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Calcium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-140-8
NACRES:
NA.21
grade:
ACS reagent
form:
solid
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

99.0-102.0% (complexometric)

form

solid

potency

2630 mg/kg LD50, skin (Rat)

impurities

≤0.003% Oxidizing substances (as NO3)
≤0.01% Insoluble matter

pH

4.5-8.5 (20 °C, 50 g/L in H2O)

mp

176 °C (dec.) (lit.)

anion traces

sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0001%
Ba: ≤0.003%
Cu: ≤0.0005%
Fe: ≤0.0003%
K: ≤0.01%
Mg: ≤0.005%
Mg and allied cations: ≤0.5%
NH4+: ≤0.005%
Na: ≤0.01%
Sr: ≤0.05%
heavy metals: ≤5 ppm

storage temp.

15-25°C

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Calcium Chloride as a Novel Stabilizer for Foot-and-Mouth Disease Virus and Its Application in the Vaccine Formulation: Demonstrates the innovative use of calcium chloride dihydrate in stabilizing foot-and-mouth disease viruses, enhancing the efficacy and stability of vaccines (Jin JS, et al., 2024).

  • Efficient regulation of cadmium accumulation by carboxymethylammonium chloride in rice: Discusses the role of calcium chloride dihydrate in the absorption and regulation of heavy metals in plants, showing potential for environmental cleanup and safer agricultural practices (Fu L, et al., 2024).

  • Simultaneous impregnation and microencapsulation of CaCl(2) using silica gel and methyl cellulose for thermal energy storage applications: Highlights the use of calcium chloride dihydrate in energy storage systems, emphasizing its ability to enhance thermal properties and energy efficiency (Shervani S, et al., 2024).

  • Microbial induced carbonate precipitation for remediation of heavy metals, ions and radioactive elements: Explores the environmental applications of calcium chloride dihydrate in bioremediation, particularly in treating water and soil contaminated with heavy metals and radioactive materials (Taharia M, et al., 2024).

  • Hybrid Polymer Salogels for Reversible Entrapment of Salt-Hydrate-Based Thermal Energy Storage Materials: Investigates the integration of calcium chloride dihydrate in polymer matrices for reversible thermal energy storage, offering advancements in materials science and energy management (Rajagopalan KK, et al., 2024).

Linkage

Replaces: CX0134; CX0130-1 CX0134-1; CX0134-3; CX0130; CX0130-6

Analysis Note

Assay (complexometric): 99.0 - 102.0 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.01 %
Acidity or alkalinity: passes test
pH-value (5 %; water, 25 °C): 4.5 - 8.5
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (ACS): ≤ 5 ppm
Al (Aluminium): ≤ 0.0001 %
Ba (Barium):
≤ 0.003 %
: passes test
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0003 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.005 %
Na (Sodium): ≤ 0.01 %
NH₄ (Ammonium): ≤ 0.005 %
Sr (Strontium): ≤ 0.05 %
Magnesium and alkali metals: ≤ 0.5 %
Oxidizing substances (as NO₃): ≤ 0.003 %
Corresponds to ACS, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination of nitrite ions in compost by Griess method using Spectroquant®

Learn about the photometric determination of nitrate ions in soil using Spectroquant® test kits and spectrophotometer.

Photometric determination of nitrite ions in soil samples by Griess method using Spectroquant® test kits and spectrophotometer.

Learn about the photometric determination of nitrate ions in compost using Spectroquant® test kits and spectrophotometer.

See All

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service