Skip to Content
Merck
All Photos(1)

Key Documents

1.05230

Sigma-Aldrich

Oil red O (C.I. 26125)

for microscopy Certistain®

Synonym(s):

Oil red O (C.I. 26125)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
408.5
MDL number:
UNSPSC Code:
41116130
EC Index Number:
215-295-3
Pricing and availability is not currently available.

composition

Dye, ≥80% spectrophotometric assay

mp

182-186 °C

bulk density

270 kg/m3

εmax

725-910 at 528-534 nm in chloroform at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C26H24N4O/c1-16-9-10-17(2)22(13-16)27-28-23-14-19(4)24(15-18(23)3)29-30-26-21-8-6-5-7-20(21)11-12-25(26)31/h5-15,29H,1-4H3/b28-27?,30-26-

InChI key

HJAYEODIXUYVIC-LTFAAIOASA-N

Looking for similar products? Visit Product Comparison Guide

General description

The dry dye Oil red O (C.I. 26125) for microscopy Certistain®, is used for the preparation of a staining solution that can be used for the detection of lipids in histological cryo sections. As a Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. Oil red O staining is used for the detection of neutral fat or polyethylene granula in e. g. histological tissues, in joint pathology or in cytological sample material. This Oil red O solution is ready-to-use and results in a brilliant, bright red staining of lipids, while the nuclei are counterstained blue with Hematoxylin solution acc. to Gill II (Cat. no. 105175).
The staining of lipids in histology specimens is a physical process. Oil red O is a lipophilic dye; due to its solubility characteristics it diffuses to the tissue lipids. It is registered as IVD and certified as CE product. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Certistain® dry dye used to detect lipids in cryosections

Analysis Note

Dye content (spectrophotometrically)≥ 80 %Identity (UV/VIS-Spectrum)passes testAbsorption maximum λ max (chloroform)528 - 534 nmSpec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; chloroform)725 - 910TLC-Testpasses testLoss on drying (110 °C)≤ 8 %Suitability for microscopypasses test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service