Skip to Content
Merck
All Photos(2)

Documents

1.04239

Sigma-Aldrich

Glutardialdehyde solution 25%

for electron microscopy

Synonym(s):

Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Beilstein:
605390
MDL number:
UNSPSC Code:
12352114
NACRES:
NA.21

vapor pressure

0.27 hPa ( 20 °C)

Quality Level

form

liquid

concentration

23.5-26.5% (alkalimetric)

technique(s)

electron microscopy: suitable

color

colorless

pH

>3.0 (20 °C in H2O)

bp

100.5 °C/1013 hPa

mp

-6 °C

density

1.06 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[H]C(CCCC([H])=O)=O

InChI

1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2

InChI key

SXRSQZLOMIGNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glutardialdehyde solution 25% - for electron microscopy, is a fixing agent for specimens destined for morphological and enzyme histochemical analysis. The 25% Glutardialdehyde solution is used for the fixation of specimen of human origin for semi-thin cuttings and electron microscopy. It is diluted with phosphate buffer(Potassium dihydrogen phosphate for analysis EMSURE® ISO (Product number 1.04873) or di-Sodium hydrogen phosphate anhydrous for analysis EMSURE® ACS,Reag. Ph Eur (Product number 1.06586)
to achieve the corresponding working concentration. Alternatively, especially sensitive material used for enzyme histochemistry, will be fixed with glutardialdehyde according to P.J. Anderson, purified and filled into ampoules under nitrogen (Product number 1.12179). Sensitive enzymes e.g. endogenous peroxidase can often only be visualized in this way. It is an IVD registered product and CE certified, thus can be used for clinical diagnostic purposes. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Assay (alkalimetric): 23.5 - 26.5 %
pH-value: ≥ 3.0
Density (d 20 °C/ 4 °C): 1.060 - 1.065
Absorbance A (235 nm; 1:200 deluted with water; 1 cm): ≤ 0.3

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service