1.02554
Cobalt(II) nitrate hexahydrate
for analysis (max. 0.001% Ni) EMSURE® ACS,Reag. Ph Eur
Synonym(s):
Cobalt(II) nitrate hexahydrate
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Agency
reag. Ph. Eur.
product line
EMSURE®
Assay
99.0-102.0% (Co(NO3)2.6H2O basis, complexometric)
form
solid
potency
691 mg/kg LD50, oral (Rat)
impurities
≤0.005% Insoluble matter
pH
4.0 (20 °C, 100 g/L in H2O)
mp
57 °C
solubility
2170 g/L
density
1.87 g/cm3 at 20 °C
bulk density
800 kg/m3
anion traces
chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%
cation traces
Ca: ≤0.005%
Cu: ≤0.001%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
Mn: ≤0.002%
NH4+: ≤0.05%
Na: ≤0.05%
Ni: ≤0.001%
Pb: ≤0.001%
Zn: ≤0.005%
storage temp.
2-30°C
InChI
1S/Co.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;
InChI key
QGUAJWGNOXCYJF-UHFFFAOYSA-N
Linkage
Analysis Note
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.005 %
Mn (Manganese): ≤ 0.002 %
Na (Sodium): ≤ 0.05 %
NH₄ (Ammonium): ≤ 0.05 %
Ni (Nickel): ≤ 0.001 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.005 %
Legal Information
Signal Word
Danger
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation
Target Organs
Lungs
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service