Skip to Content
Merck
All Photos(1)

Key Documents

1.01800

Millipore

Alkaline Peptone Water

According to ISO 8914, FDA-BAM, APHA, and AOAC; For microbiology

Synonym(s):

APW; Peptone Water, alkaline; Peptone Water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

AOAC®
APHA
FDA-BAM
ISO 8914

Quality Level

sterility

non-sterile

form

granular

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

pH

8.3-8.7 (25 °C, 20 g/L in H2O, after autoclaving)

solubility

20 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Vibrio spp.

General description

Alkaline Peptone Water is recommended for the enrichment and isolation of Vibrio cholerae and Vibrio species. The growth of a broad spectrum of Vibrio sp. is promoted by peptones, a sodium chloride concentration of 10 g/liter, and a high pH of 8.5. The alkaline pH of this medium allows the growth of Vibrio species, while it inhibits the growth of other unwanted flora.

Application

Alkaline Peptone Water is used for the enrichment of Vibrio cholerae and Vibrio sp. in food and other materials.

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellow
pH-value (25 °C): 8.3 - 8.7
Colony count (Vibrio vulnificus ATCC 33149): ≥ 1E+06 CFU/ml
Colony count (Vibrio cholerae Eltor Inaba CH 38): ≥ 1E+06 CFU/ml
Colony count (Vibrio cholerae Eltor Ogawa CH 60): ≥ 1E+06 CFU/ml
Colony count (Vibrio parahaemolyticus ATCC 17802 (WDCM 00037)): ≥ 1E+06 CFU/ml
Incubation: 24 hrs.; 35 °C; aerobic.
Reference: bloog agar.

Legal Information

AOAC is a registered trademark of AOAC International
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service