Skip to Content
Merck
All Photos(1)

Documents

1.01582

Supelco

Tetrachloroauric(III) acid trihydrate

99%, for analysis EMSURE®

Synonym(s):

Tetrachloroauric(III) acid trihydrate 99%, Gold(III)chloride hydrochloric acid, Gold trichloride hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HAuCl4 · 3H2O
CAS Number:
Molecular Weight:
393.83
MDL number:
UNSPSC Code:
12352301
EC Index Number:
240-948-4
NACRES:
NA.21

product name

Tetrachloroauric(III) acid trihydrate 99%, for analysis EMSURE®

Quality Level

product line

EMSURE®

form

solid

mp

30 °C

density

3.9 g/cm3 at 20 °C

storage temp.

no temp limit

InChI

1S/Au.4Cl/q-1;;;;/p+1

InChI key

UZPNXZZKOCLVOD-UHFFFAOYSA-O

Analysis Note

Identity (Raman-spectrum): conforms
Assay (AuCl4H, calculated of Au): 99 - 100 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service