Skip to Content
Merck
All Photos(3)

Documents

398470

Sigma-Aldrich

Gold(III) bromide

99.9% trace metals basis

Synonym(s):

Auric bromide, Gold tribromide, Gold(3+) tribromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AuBr3
CAS Number:
Molecular Weight:
436.68
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: gold

impurities

≤1500.0 ppm Trace Metal Analysis

solubility

H2O: slightly soluble(lit.)
ethanol: slightly soluble(lit.)

SMILES string

Br[Au](Br)Br

InChI

1S/Au.3BrH/h;3*1H/q+3;;;/p-3

InChI key

OVWPJGBVJCTEBJ-UHFFFAOYSA-K

General description

Gold(III) bromide, also known as auric bromide, serves as a starting material in the productionof gold nanoparticles, catalysts, and various organogold compounds.

Application

Gold(III) bromide can be used:
  • As a precursor to synthesize gold nanoparticles for perovskite nanocrystals.
  • As a catalyst for dehydrogenative coupling of amines and phenylglyoxal derivatives to synthesize α-Ketoamide.
  • For the chemoselective annulation of isocyanates with 2H-azirine.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mild gold-catalyzed aerobic dehydrogenative coupling of amines and phenylglyoxal derivatives
Ying Shao, et al.},
Journal of Organometallic Chemistry, 767, 60-64}-60-64} (2014)
AuBr3-Catalyzed Chemoselective Annulation of Isocyanates with 2H-Azirine
Yufeng Wu, et al.
Chemistry?A European Journal , 29, 4093-4099 (2019)
Au-CsPbBr3 hybrid architecture: anchoring gold nanoparticles on cubic perovskite nanocrystals
Subila K Balakrishnan and Prashant V Kamat
ACS Energy Letters, 2, 88-93 (2017)

Articles

We are proud to offer a treasure-trove of gold precatalysts and silver salts, as well as an extensive portfolio of unsaturated building blocks to accelerate your research success in this exciting field.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service