Skip to Content
Merck
Ethanol API Grade

Ethanol

Ethanol API Grade
Synonym(s):
Ethyl alcohol, Ethanol
Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique