Skip to Content
Merck
All Photos(1)

Documents

1.00579

Sigma-Aldrich

DPX new

non-aqueous mounting medium for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NA.21

Quality Level

IVD

for in vitro diagnostic use

color

colorless

refractive index

n/D 1.519

transition temp

flash point 22.0 °C

density

0.9068 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

DPX new is a synthetic non-aqueous mounting medium for Microscopy. In its formulation, the teratogenic plasticizer Dibutyl phthalate (DBP) has been avoided, which renders the composition safer for environment and more user-friendly than the conventionally used mounting agent DPX. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Refractive index (20°C) 1.518 - 1.521
Viscosity (20°C) 600 - 700 mPa*s

Application

Non-aqueous mounting medium

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amirah Baharin et al.
Journal of research in medical sciences : the official journal of Isfahan University of Medical Sciences, 25, 20-20 (2020-03-17)
Previous studies have shown that morphine negatively effects male fertility while Phoenix dactylifera (dates) could cure male infertility by the exhibition of antagonist effects. This study was conducted to assess the possible ameliorating effects of dates on the histological features
Diana Furcila et al.
Frontiers in neuroanatomy, 13, 99-99 (2020-01-11)
A variety of anatomical alterations have been reported in the hippocampal formation of patients with Alzheimer's Disease (AD) and these alterations have been correlated with cognitive symptoms in the early stages of the disease. Major hallmarks in AD are the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service