Skip to Content
Merck
All Photos(1)

Key Documents

1.00414

Millipore

Presence-Absence Broth

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

form

medium granules (dehydrated (DCM))

Quality Level

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Escherichia coli

General description

for microbiology

Analysis Note

Appearance (clearness): clear
Appearance (colour): violet
pH-value (25 °C): 6.6 - 7.0
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Escherichia coli ATCC 11775 (WDCM 00090)): good to very good
Growth (Enterococcus faecalis ATCC 29212 (WDCM 00087)): fair
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): fair to good
Colour change to yellow (Escherichia coli ATCC 25922 (WDCM 00013)): +
Colour change to yellow (Escherichia coli ATCC 11775 (WDCM 00090)): +
Colour change to yellow (Enterococcus faecalis ATCC 29212 (WDCM 00087)): none to poor
Colour change to yellow (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): -
Gas (Escherichia coli ATCC 25922 (WDCM 00013)): +
Gas (Escherichia coli ATCC 11775 (WDCM 00090)): +
Gas (Enterococcus faecalis ATCC 29212 (WDCM 00087)): -
Gas (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): -
Incubation: 24-48 hrs; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service