Skip to Content
Merck
All Photos(2)

Key Documents

1.00244

Supelco

Citric acid monohydrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2 · H2O
CAS Number:
Molecular Weight:
210.14
Beilstein:
4018641
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-069-1
NACRES:
NA.21
Assay:
99.5-100.5% anhydrous basis (acidimetric)
grade:
ACS reagent
form:
solid
solubility:
880 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor pressure

<1 Pa ( 25 °C)

product line

EMSURE®

Assay

99.5-100.5% anhydrous basis (acidimetric)

form

solid

potency

3000 mg/kg LD50, oral (Rat)

composition

Water, 7.5-8.8% Karl Fischer

impurities

≤200 ppm Sulfated ash
≤50 ppm In water insoluble matter

color

clear
white to almost white

pH

1.85 (25 °C, 50 g/L in H2O)

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

mp

135-152 °C

transition temp

flash point 173.9 °C

solubility

880 g/L

density

1.54 g/cm3 at 20 °C

bulk density

800‑1000 kg/m3

anion traces

chloride (Cl-): ≤5 ppm
oxalate (as oxalic acid): ≤50 ppm
phosphate (PO43-): ≤10 ppm
sulfate (SO42-): ≤20 ppm

cation traces

Al: ≤0.2 ppm
As: ≤1 ppm
Cu: ≤5 ppm
Fe: ≤3 ppm
Hg: ≤1 ppm
Pb: ≤0.5 ppm
heavy metals (as Pb): ≤5 ppm

storage temp.

2-30°C

SMILES string

OC(CC(O)(C(O)=O)CC(O)=O)=O.O

InChI

1S/C6H8O7.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1H2

InChI key

YASYEJJMZJALEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Biopolymeric Blends of Thermoplastic Starch and Polylactide as Sustainable Packaging Materials: This publication uses citric acid monohydrate in the synthesis of biodegradable polymers, highlighting its utility in developing sustainable materials within the industrial manufacturing and environmental sectors (Jozinović A et al., 2024).

Linkage

Replaces: CX1725-1; CX1725-3; CX1725

Analysis Note

Assay (acidimetric, calc. on anhydrous substance): 99.5 - 100.5 %
Identity (IR-spectrum): passes test
Appearance: white to almost white or colorless crystals
Appearance of solution (200 g/l; water): clear (≤ 3 NTU) and not more intense in color than reference solution Y₇, BY₇ or GY₇
In water insoluble matter: ≤ 50 ppm
Chloride (Cl): ≤ 5 ppm
Phosphate (PO₄): ≤ 10 ppm
Sulphate (SO₄): ≤ 20 ppm
Heavy metals (as Pb): ≤ 5 ppm
Al (Aluminium): ≤ 0.2 ppm
As (Arsenic): ≤ 1 ppm
Hg (Mercury): ≤ 1 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 3 ppm
Fe (Iron, acc. to Reag. Ph  Eur.): suitable to use in the limit test for iron
Pb (Lead): ≤ 0.5 ppm
Oxalates (as C₂H₂O₄): ≤ 50 ppm
Readily carbonisable substance: passes test
Sulfated ash: ≤ 200 ppm
Water (according to Karl Fischer): 7.5 - 8.8 %
Corresponds to ACS, ISO 6353/2, Reag. Ph. Eur.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

345.0 °F - closed cup

Flash Point(C)

173.9 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service