Skip to Content
Merck
All Photos(3)

Key Documents

27109

Sigma-Aldrich

Citric acid

meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Beilstein:
782061
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
39021801
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

99.5-100.5% (based on anhydrous substance)

form

grit

quality

anhydrous
meets analytical specification of Ph. Eur., BP, USP, E330

expl. lim.

8 %, 65 °F

impurities

residual solvents, complies
≤0.0005% heavy metals (as Pb)
≤0.5% water (Karl Fischer)

ign. residue

≤0.05% (as SO4)

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

mp

153-159 °C (lit.)

solubility

water: 383 g/L at 25 °C

anion traces

chloride (Cl-): ≤50 ppm
oxalate (as oxalic acid): ≤100 ppm
sulfate (SO42-): ≤100 ppm

cation traces

As: ≤1 ppm
Ca: ≤200 ppm
Cu: ≤10 ppm
Fe: ≤10 ppm
Hg: ≤1 ppm
Pb: ≤0.5 ppm
Zn: ≤10 ppm

suitability

complies for appearance of solution
complies for reaction against H2SO4

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

Application

Citric acid was used as a test reagent for studying whether the novel pyridazinylpiperazine analog TPRV-1 inhibitor can reduce cough reflex induced by citric acid and capsaicin in guinea pig.[1]

Biochem/physiol Actions

Citric acid in dietary form can augments absorption of aluminium in antacids.[2] It also facilitates the phytoremediation of heavy metal contaminated soil and can transform cadmium into more transportable forms.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Slanina et al.
Clinical chemistry, 32(3), 539-541 (1986-03-01)
Ten healthy men ingested, twice daily between meals, during each of the seven-day experimental periods: (a) citric acid (as lemon juice), (b) Al(OH)3, or (c) Al(OH)3 + citric acid. Whole blood sampled after each dietary period was analyzed electrothermally after
Sum Yee Leung et al.
Cough (London, England), 3, 10-10 (2007-12-25)
Cough reflex can be induced by the pepper extract capsaicin and by low pH in guinea-pig airways. Transient receptor potential vanniloid-1 (TPRV-1) is expressed in the sensory and afferent nerve fibres in airways. We hypothesized that a novel pyridazinylpiperazine analog
Y X Chen et al.
Chemosphere, 50(6), 807-811 (2003-04-12)
Adsorption and hydroponics experiments were conducted to study the role of citric acid on the phytoremediation of heavy metal contaminated soil. The results show that addition of citric acid decreased the adsorption of both lead and cadmium, such an effect
Maria José Garcia-Fernandez et al.
Biofouling, 29(3), 261-271 (2013-02-27)
Prevention and management of wound infections receive a lot of attention, since the presence of micro-organisms interferes with the wound-healing process. The aim of this work was to use cyclodextrins (CDs) to endow hydrogels and gauzes with the ability to
Benjamin Nota et al.
American journal of human genetics, 92(4), 627-631 (2013-04-09)
The Krebs cycle is of fundamental importance for the generation of the energetic and molecular needs of both prokaryotic and eukaryotic cells. Both enantiomers of metabolite 2-hydroxyglutarate are directly linked to this pivotal biochemical pathway and are found elevated not

Questions

  1. Regarding Citric Acid (Product Number: 27109, Batch Number: STBL2004), confirm if it is a primary or secondary standard. Additionally, if it is a secondary standard, confirm the batch number of the primary traceability standard.

    1 answer
    1. This product is not considered an analytical standard. It meets the analytical specifications of the European, British, and United States Pharmacopeia, but is neither a primary nor secondary standard. It is intended for use in general research applications. See the links below to review additional products that may be of interest:

      1134369, USP reference standard
      https://www.sigmaaldrich.com/product/usp/1134368

      A1202000, EP reference standard
      https://www.sigmaaldrich.com/product/sial/a1202000

      94676, Secondary reference standard, traceable to NIST/NMIJ
      https://www.sigmaaldrich.com/product/sial/94676

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service