Skip to Content
Merck
All Photos(2)

Key Documents

06-340

Sigma-Aldrich

Anti-Myc Antibody

Upstate®, from rabbit

Synonym(s):

Transcription factor p64, avian myelocytomatosis viral oncogene homolog, myc proto-oncogene protein, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, Xenopus, avian

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... MYC(4609)

General description

c-Myc-, N-Myc- and L-Myc-encoded proteins function in cell proliferation, differentiation and neoplastic disease. Myc proteins are nuclear proteins with relatively short half lives. Amplification of the c-Myc gene has been found in several types of human tumors including lung, breast and colon carcinomas, while the N-Myc gene has been found amplified in neuroblastomas. The L-Myc gene has been reported to be amplified and expressed at high level in human small cell lung carcinomas. The c-Myc protein is a transcription factor, which is encoded by the c-Myc gene on human chromosome 8q24. c-Myc is commonly activated in a variety of tumor cells and plays an important role in cellular proliferation, differentiation, apoptosis and cell cycle progression.

Specificity

Recognizes Myc isoforms Mr 50-67kDa.
Will also cross-react with avian and Xenopus.

Immunogen

Bacterially expressed fusion-protein corresponding to the full-length human Myc.

Application

Immunoprecipitation:
4 µg of a previous antibody immuno-precipitated Myc as shown by an independent laboratory.

Immunocytochemistry:
This antibody has been used for immunocytochemistry by an independent laboratory. (Ohtsuka, T., 1998)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-Myc Antibody (rabbit polyclonal antibody) validated in ChIP, ICC, IP, WB to detect Myc also known as Transcription factor p64, avian myelocytomatosis viral oncogene homolog, myc proto-oncogene protein.

Quality

Routinely evaluated by western blot on A431 lysates from EGF-stimulated human A431 cells, human A431 cells and HeLa nuclear extract.

Western Blot Analysis:
0.5-2 µg/mL of this lot detected Myc in A431 lysates from EGF-stimulated human A431 cells. 0.5-2 µg/mL of a previous lot detected Myc in RIPA lysates from human A431 cells and HeLa nuclear extract.

Target description

67 kDa

Physical form

Format: Purified
Immunoaffinity Chromatography
Purified rabbit polyclonal IgG in buffer containing PBS, 0.05% sodium azide with 2 mg/mL BSA. Frozen solution.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Positive Antigen Control: Catalog #12-302, EGF-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

c-Myc activation by Theileria parasites promotes survival of infected B-lymphocytes.
Dessauge, F; Hilaly, S; Baumgartner, M; Blumen, B; Werling, D; Langsley, G
Oncogene null
Proteomic profiling of Myc-associated proteins.
Agrawal, P; Yu, K; Salomon, AR; Sedivy, JM
Cell Cycle null
Tina Stoelzle et al.
BMC biology, 7, 63-63 (2009-09-30)
The oncoprotein c-Myc has been intensely studied in breast cancer and mouse mammary tumor models, but relatively little is known about the normal physiological role of c-Myc in the mammary gland. Here we investigated functions of c-Myc during mouse mammary
Lee A Campbell et al.
The FEBS journal, 284(23), 4096-4114 (2017-11-09)
Microglia, the resident macrophages of the brain, play a key role in the pathogenesis of HIV-associated neurocognitive disorders (HAND) due to their productive infection by HIV. This results in the release of neurotoxic viral proteins and pro-inflammatory compounds which negatively
T-cell lymphomas mask slower developing B-lymphoid and myeloid tumours in transgenic mice with broad haemopoietic expression of MYC.
Smith, DP; Bath, ML; Harris, AW; Cory, S
Oncogene null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service