Skip to Content
Merck
All Photos(1)

Key Documents

SAE0201

Sigma-Aldrich

Anti-C-Myc Magnetic Beads

Magnetic agarose, suspension

Synonym(s):

Anti-c-Myc Tag antibody produced in rabbit, Myc Tag Antibody, c- Myc epitope Tag

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$395.00

$395.00


Estimated to ship onApril 03, 2025



Select a Size

Change View
1 ML
$395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.56

$395.00


Estimated to ship onApril 03, 2025


conjugate

magnetic beads conjugate

Quality Level

form

suspension

packaging

pkg of 1 mL

shipped in

wet ice

storage temp.

2-8°C

General description

This product is a conjugate of activated magnetic cross-linked agarose beads to a polyclonal antibody that was raised against the c-Myc sequence. The c-Myc peptide tag sequence (EQKLISEEDL) is an epitope that is located within amino acid residues 410-419 of the human c-Myc protein. This tag is used in recombinant protein expression and for purification of c-Myc tagged proteins expressed in bacterial and mammalian cells.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Dear Sir, Madam, What is the size of the bead please? Is it possible to seperate cells rather than proteins in a lysate? Best,

    1 answer
    1. The particle size has not been determined for this product. The product has been optimized for use in protein purification in which the cells would be lysed prior to applying a magnetic field to achieve separation. In principal, there is no reason that the product could not be used in cell separation, however the it has not been tested as such. The researcher would have to determine suitability.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service