Skip to Content
Merck
All Photos(3)

Key Documents

06-1074

Sigma-Aldrich

Anti-Interleukin 21 (IL-21) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Za11, IL-21, IL21, Interleukin-21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... IL21(59067)

General description

Interleukin 21 (IL-21) is a cytokine that has immunoregulatory activity on cells of the immune system, including natural killer (NK) cells and cytotoxic T cells that can destroy virally infected or cancerous cells. IL-21 induces cell division/proliferation in its target cells and is expressed in activated human CD4+ T cells. IL-21 may promote the transition between innate and adaptive immunity and may play a role in proliferation and maturation of natural killer (NK) cells in synergy with IL15. IL-21 expression is up-regulated in Th2 and Th17 subsets of T helper cells, as well as follicular T cells. Furthermore IL-21 is expressed in NK T cells regulating the function of these cells. IL-21 is also produced by Hodgkin′s lymphoma (HL) cancer cells and may explain a great deal of the behavior of classical Hodgkin′s lymphoma including clusters of other immune cells gathered around HL cells in cultures.

Specificity

Cow, canine, equine, and porcine (93% sequence homology). Rat (86% sequence homology).
This antibody recognizes the C-terminus of IL-21.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human IL-21.

Application

Anti-Interleukin 21 (IL-21) Antibody is an antibody against Interleukin 21 (IL-21) for use in FC, IH(P) & WB.
Flow Cytometry Analysis: 5 µg from a previous lot detected IL-21 in human PBMC′s.

Immunohistochemistry Analysis: A 1:100 dilution from a previous lot detected IL-21 in mouse spleen tissue.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Quality

Evaluated by Western Blot in K562 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected IL-21 on 10 µg of K562 cell lysate.

Target description

A molecular weight of 18 kDa was observed; however, UniProtKB/Swiss-Prot entry Q9HBE4 (IL21_HUMAN) describes 2 isoforms produced by alternative splicing: isoform 1 at 18 kDa, isoform 2 at 17 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in PBS with 0.06% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maha Zohra Ladjemi et al.
American journal of respiratory and critical care medicine, 199(5), 592-602 (2018-10-20)
Accumulation of B cells and lymphoid follicles (LFs) has been described in chronic obstructive pulmonary disease (COPD) airways, but the functional status of lung B cells remains poorly known. To characterize LFs for expression of IgA, the main mucosal antibody.
Aditya Rayasam et al.
Journal of neuroinflammation, 19(1), 125-125 (2022-05-28)
Ischemic stroke is a leading cause of mortality worldwide, largely due to the inflammatory response to brain ischemia during post-stroke reperfusion. Despite ongoing intensive research, there have not been any clinically approved drugs targeting the inflammatory component to stroke. Preclinical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service